Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium isopropoxide, pure

Catalog No. AC301240250
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle

CAS: 2388-10-5 | C3H7LiO | 66.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2388-10-5
Molecular Formula C3H7LiO
Molecular Weight (g/mol) 66.03
MDL Number MFCD00048286
InChI Key HAUKUGBTJXWQMF-UHFFFAOYSA-N
Synonym lithium isopropoxide, lithium propan-2-olate, lithium isopropanolate, lithium 2-propanolate, acmc-1ch0u, lithium isopropoxide solution, 2-propanol, lithiumsalt 1:1, lithium 1+ propan-2-olate
PubChem CID 23673350
IUPAC Name lithium;propan-2-olate
SMILES [Li+].CC(C)[O-]

Specifications

Color Beige to Yellow or Tan
Density 0.69
Infrared Spectrum Authentic (nujol)
Packaging Glass bottle
Linear Formula LiOCH(CH3)2
Quantity 25 g
Solubility Information Solubility in water: reacts
Formula Weight 66.03
Percent Purity ≥94% (corrected for LiOH+Li2CO3)
Grade Pure
Physical Form Powder
Chemical Name or Material Lithium isopropoxide
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Flammable solid.
Causes severe skin burns and eye damage.
Reacts violently with water.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.