Learn More
Lithium methoxide, 98+%
Description
Lithium methoxide is used in sol-gel synthesis of lithium borophosphates. It is also useful for deprotonation reactions in organic synthesis. Further, it serves as a mild base in organic synthesis especially in transesterifications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | >500°C (decomposition) |
| Color | White |
| Linear Formula | LiOCH3 |
| Quantity | 25 g |
| UN Number | UN3206 |
| Beilstein | 3552389 |
| Sensitivity | Air and moisture sensitive |
| Solubility Information | Soluble in methanol. |
| IUPAC Name | lithium(1+) methanolate |
| Formula Weight | 37.98 |
| Show More |
Safety and Handling
| Hazard Category | H314-X |
| Hazard Statement | GHS H Statement H251-H314 Self-heating: may catch fire. Causes severe skin burns and eye damage. |
| Precautionary Statement | P235+P410-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
| DOTInformation | Transport Hazard Class: 4.2; Packing Group: II; Proper Shipping Name: ALKALI METAL ALCOHOLATES, SELF-HEATING, CORROSIVE, N.O.S. |
| EINECSNumber | 212-737-7 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.