Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium sulfide, 99.9% (metals basis)

Catalog No. AA1283909
Click to view available options
Quantity:
2 g
10 g
50 g
250 g

CAS: 12136-58-2 | Li2S | 45.94 g/mol

It act as a superconductor and widely used in lithium-sulfur batteries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12136-58-2
Molecular Formula Li2S
Molecular Weight (g/mol) 45.94
MDL Number MFCD00011085
InChI Key GLNWILHOFOBOFD-UHFFFAOYSA-N
Synonym lithium sulfide, lithium sulfide li2s, lithium sulphide, anhydrous, li2s, zwdbutfcwlvlcq-uhfffaoysa-n, lithium sulfide trace metals basis
PubChem CID 64734
SMILES [Li+].[Li+].[S--]

Specifications

Melting Point 900°C to 975°C
Color Yellow
Physical Form Powder
Quantity 10 g
Assay Percent Range 99.9% (Metal basis)
Solubility Information soluble in water and ethanol
IUPAC Name dilithium(1+) sulfanediide
Formula Weight 45.95
Odor Like rotten eggs
Sensitivity Moisture sensitive
Appearance Yellow powder
Density 1.66 g/cm3 at 20°C
Chemical Name or Material Lithium sulfide
Show More Show Less
How should Lithium sulfide, 99.9% (metals basis) (Cat. No. 012839.xx) be stored and handled?

Lithium sulfide releases flammable gas when in contact with water and should be kept away from water, moist air, or acids. Lithium sulfide should be stored and handled under a dry, inert gas atmosphere. Store at ambient temperatures, typically 20-25 degrees C.

Hazard Category H301-H314-H335
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation DOT Class: 8; Packaging Group: II
EINECSNumber 235-228-1
RTECSNumber OJ6439500
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.