Learn More
Lithium thiocyanate hydrate
Description
Lithium thiocyanate hydrate is used as solution in infra red ultrasonic technique and also used as solvent for the fibers of mulberry in silk production process. It is used to determined the molecular structure of benzo-13-crown-4 compound.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Physical Form | Crystalline |
| Quantity | 50 g |
| Solubility Information | Soluble in water. |
| Formula Weight | 65.02 (Anhydrous) |
| Sensitivity | Hygroscopic |
| Chemical Name or Material | Lithium thiocyanate hydrate |
Safety and Handling
| Hazard Category | H302+H312+H332 |
| Hazard Statement | GHS H Statement H302-H312-H332 Harmful if swallowed. Harmful in contact with skin. Harmful if inhaled. |
| Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c |
| EINECSNumber | 209-135-1 |
| RTECSNumber | XL1540000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.