Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium titanium oxide

Catalog No. p-4781977
Click to view available options
Quantity:
250 g

CAS: 12031-82-2 | Li2O3Ti | 109.74 g/mol

Lithium titanium oxide is used in battery, porcelain enamels and ceramic insulating bodies. It is frequently utilized as a flux due to its good stability. It is used as a cathode in layer one of a double layer cathode for molten carbonate fuel cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12031-82-2
Molecular Formula Li2O3Ti
Molecular Weight (g/mol) 109.74
MDL Number MFCD00016181
InChI Key AXQWGBXVDWYWDE-UHFFFAOYSA-N
Synonym lithium titanate, lithium titanium oxide, dilithium dioxido oxo titanium, 2li.o3ti, dilithium 1+ ion oxotitaniumbis olate, dilithium 1+ oxotitaniumbis olate
IUPAC Name titanium(4+) dilithium(1+) trioxidandiide
SMILES [Li+].[Li+].[O--].[O--].[O--].[Ti+4]

Specifications

Melting Point 1520°C to 1564°C
Color White
Physical Form Powder
Quantity 250 g
Solubility Information Insoluble in water.
Formula Weight 109.78
Odor Odorless
Chemical Name or Material Lithium titanium oxide
EINECSNumber 234-759-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.