Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LIVE/DEAD™ Fixable Near IR (876) Viability Kit, for 808 nm excitation

Catalog No. p-7230013
Encompass_Preferred
Click to view available options
Quantity:
200 Assays
400 Assays
80 Assays

The LIVE/DEAD Fixable viability kits allow for clear determination of live and dead cells, enabling more precise analysis with both conventional and spectral flow cytometry.

  • Fixable viability kits allow for clear determination of live and dead cells, enabling more precise analysis with both conventional and spectral flow cytometry
  • Cells labeled with LIVE/DEAD Fixable viability dyes can be washed, fixed, permeabilized, and stained for intracellular antigens while maintaining distinct live and dead cell populations
  • The narrow and unique emission spectra are ideal for expanding high-parameter flow cytometry experiments
  • Designed to provide a bright signal with minimal overlap into adjacent channels, simplifying spectral compensation
  • Since live and dead cells can be discriminated using a single flow cytometer channel, these viability dyes are an ideal choice for multi-color experiments.
  • Their unique spectral fingerprints are a valuable addition to spectral flow cytometry experiments
  • Dyes have been conveniently packaged in single-use vials to help ensure dye stability and performance over time
  • Dissolve the dye in the included anhydrous DMSO and use immediately, or store at -20°C, avoiding freeze-thaw cycles
  • The LIVE/DEAD Fixable Dead Cell Stain Sampler Kit provides one vial (40 assays) of each of eight available dead cell stains
TRUSTED_SUSTAINABILITY

Specifications

Quantity 80 Assays
Description LIVE/DEAD™ Fixable Near IR (876) Viability Kit, for 808 nm excitation
Detection Method Fluorescence
Conjugate LIVE/DEAD Fixable Near IR (876) Cell Stain
For Use With (Application) Flow Cytometry
Product Type Cell Viability Kit
Format Tubes
Color Near-IR
Content And Storage Contains 2 vials of LIVE/DEAD fixable dead cell stain and 500 μL DMSO.

Store at -20°C.
Assay Viability Assay
Compatible Cells Eukaryotic Cells
Excitation/Emission 849/876 nm
Flow Cytometer Laser Lines 808 nm
Solubility DMSO (Dimethylsulfoxide)
Shipping Condition Room Temperature
Product Line LIVE/DEAD™
For Use With (Equipment) Flow Cytometer
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.