Learn More
Description
Specifications
Specifications
| Antigen | Livin |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000-1:2000, Immunohistochemistry 1:1000-1:5000, Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000 |
| Formulation | Neat whole antisera with 0.05% Sodium Azide |
| Gene Accession No. | Q96CA5 |
| Gene Alias | baculoviral IAP repeat containing 7, baculoviral IAP repeat-containing 7, KIAPRING finger protein 50, Kidney inhibitor of apoptosis protein, Livin, livin inhibitor-of-apoptosis, Melanoma inhibitor of apoptosis protein, MLIAPlivin inhibitor of apoptosis, ML-IAPmliap, RNF50LIVINbaculoviral IAP repeat-containing protein 7 |
| Gene Symbols | BIRC7 |
| Host Species | Rabbit |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
