Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Livin Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 5 publications

$243.50 - $353.50

Specifications

Antigen Livin
Dilution Western Blot 1:1000-1:2000, Immunohistochemistry 1:1000-1:5000, Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000
Applications Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB10056145T
SDP
View Documents
Novus Biologicals
NB10056145SS
0.025 mL
Each for $243.50
Only null left
Add to Cart
 
NB10056145
SDP
View Documents
Novus Biologicals
NB10056145
0.05 mL
Each for $353.50
Only null left
Add to Cart
 
Description

Description

Livin Polyclonal specifically detects Livin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications

Specifications

Livin
Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Apoptosis, Zinc Finger
Neat whole antisera with 0.05% Sodium Azide
baculoviral IAP repeat containing 7, baculoviral IAP repeat-containing 7, KIAPRING finger protein 50, Kidney inhibitor of apoptosis protein, Livin, livin inhibitor-of-apoptosis, Melanoma inhibitor of apoptosis protein, MLIAPlivin inhibitor of apoptosis, ML-IAPmliap, RNF50LIVINbaculoviral IAP repeat-containing protein 7
BIRC7
IgG
Unpurified
Livin is a short-lived protein that is thought to be degraded by the ubiquitin-proteasome pathway. Thus users may want to do a time course when examining the effects of treatments on Livin expression. Users are encouraged to refer to the literature for additional information; for example, Ma et al. 2006. 2.
Western Blot 1:1000-1:2000, Immunohistochemistry 1:1000-1:5000, Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000
Polyclonal
Antisera
RUO
Human, Mouse, Rat
Q96CA5
79444
A synthetic peptide corresponding to amino acids 180-230 of human Livin, isoform alpha and isoform beta was used as immunogen (NP_647478).
Primary
Store at -20C. Avoid freeze-thaw cycles.
33 kDa
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.