Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Livin Antibody (88C570), HRP, Novus Biologicals™
SDP

Catalog No. NB10056548H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056548H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10056548H Supplier Novus Biologicals Supplier No. NB10056548H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Livin Monoclonal specifically detects Livin in Human samples. It is validated for Immunocytochemistry/Immunofluorescence, Western Blot.

Specifications

Antigen Livin
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 88C570
Conjugate HRP
Dilution Western Blot
Gene Accession No. Q96CA5
Gene Alias baculoviral IAP repeat containing 7, baculoviral IAP repeat-containing 7, KIAPRING finger protein 50, Kidney inhibitor of apoptosis protein, Livin, livin inhibitor-of-apoptosis, Melanoma inhibitor of apoptosis protein, MLIAPlivin inhibitor of apoptosis, ML-IAPmliap, RNF50LIVINbaculoviral IAP repeat-containing protein 7
Gene Symbols BIRC7
Host Species Mouse
Immunogen Recombinant human livin protein was used as immunogen.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 79444
Test Specificity The Livin antibody, clone 88C570 has been shown to recognize both Livin alpha (∼39 kDa) and Livin beta (∼37 kDa) isoforms (Nachmias et al, 2003). 2. The antibody also recognizes the 30 kDa (p30-Livin alpha) and 28 kDa (p28-Livin beta) apoptotic cleavage fragments of Livin alpha and beta, respectively (Nachmias et al, 2003). Apoptotic cleavage fragments of Livin have also been referred to as truncated livin (tLivin) (Nachmias et al, 2007). 3. The specificity of the antibody has been validated using Livin shRNA (Quiping et al, 2004).
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.