Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Livin Antibody (88C570), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. N233225J549 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N233225J549 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N233225J549 Supplier Novus Biologicals Supplier No. NBP233225JF549
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Livin Monoclonal specifically detects Livin in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Livin
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 88C570
Conjugate Janelia Fluor 549
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias baculoviral IAP repeat containing 7, baculoviral IAP repeat-containing 7, KIAPRING finger protein 50, Kidney inhibitor of apoptosis protein, Livin, livin inhibitor-of-apoptosis, Melanoma inhibitor of apoptosis protein, MLIAPlivin inhibitor of apoptosis, ML-IAPmliap, RNF50LIVINbaculoviral IAP repeat-containing protein 7
Gene Symbols BIRC7
Host Species Mouse
Immunogen Recombinant human livin protein was used as immunogen.
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Apoptosis, Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 79444.0
Test Specificity The Livin antibody, clone 88C570 has been shown to recognize both Livin alpha (∽39 kDa) and Livin beta (∽37 kDa) isoforms (Nachmias et al, 2003). 2. The antibody also recognizes the 30 kDa (p30-Livin alpha) and 28 kDa (p28-Livin beta) apoptotic cleavage fragments of Livin alpha and beta, respectively (Nachmias et al, 2003). Apoptotic cleavage fragments of Livin have also been referred to as truncated livin (tLivin) (Nachmias et al, 2007). 3. The specificity of the antibody has been validated using Livin shRNA (Quiping et al, 2004).
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.