Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LIX1 Antibody (RM0282-4F47), DyLight 488, Novus Biologicals™
SDP

Catalog No. NBP211779G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP211779G 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP211779G Supplier Novus Biologicals Supplier No. NBP211779G
Add to Cart
Add to Cart

Rat Monoclonal Antibody

LIX1 Monoclonal specifically detects LIX1 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen LIX1
Applications Western Blot
Classification Monoclonal
Clone RM0282-4F47
Conjugate DyLight 488
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias C5orf11, chromosome 5 open reading frame 11, FLJ25534, limb expression 1, Lix1 homolog (chicken), Lix1 homolog (mouse), protein limb expression 1 homolog
Gene Symbols LIX1
Host Species Rat
Immunogen Mouse recombinant LIX1
Purification Method Protein A or G purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 167410
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.