Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LIX1 Antibody (RM0282-4F47), PE, Novus Biologicals™
SDP

Catalog No. NB344362 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB344362 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB344362 Supplier Novus Biologicals Supplier No. NBP211779PE
Add to Cart
Add to Cart

Rat Monoclonal Antibody

LIX1 Monoclonal antibody specifically detects LIX1 in Mouse samples. It is validated for Western Blot, Neutralization

Specifications

Antigen LIX1
Applications Western Blot, Neutralization
Classification Monoclonal
Clone RM0282-4F47
Conjugate PE
Dilution Western Blot, Block/Neutralize
Formulation PBS
Gene Alias C5orf11, chromosome 5 open reading frame 11, FLJ25534, limb expression 1, Lix1 homolog (chicken), Lix1 homolog (mouse), protein limb expression 1 homolog
Host Species Rat
Immunogen Mouse recombinant LIX1
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 167410
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.