Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LKAAEAR1 Antibody (Z36P2C1*E7), DyLight 550, Novus Biologicals™
SDP

Catalog No. NBP250477R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250477R 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP250477R Supplier Novus Biologicals Supplier No. NBP250477R
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

LKAAEAR1 Monoclonal specifically detects LKAAEAR1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.

Specifications

Antigen C20orf201
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
Clone Z36P2C1*E7
Conjugate DyLight 550
Dilution Western Blot, ELISA, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias bA299N6.3, chromosome 20 open reading frame 201, hypothetical protein LOC198437, LKAAEAR motif containing 1, LKAAEAR1
Gene Symbols LKAAEAR1
Host Species Mouse
Immunogen Peptide Sequence - AIRLELFLPP (immunogen peptide is identical in mouse, rat, sheep, bovine and equine)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 198437
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.