Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LKAAEAR1 Antibody (Z36P2C1*E7), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB309359 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB309359 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB309359 Supplier Novus Biologicals Supplier No. NBP250477MFV450
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

LKAAEAR1 Monoclonal antibody specifically detects LKAAEAR1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry

Specifications

Antigen LKAAEAR1
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
Clone Z36P2C1*E7
Conjugate mFluor Violet 450 SE
Dilution Western Blot, ELISA, Immunohistochemistry
Formulation 50mM Sodium Borate
Gene Alias bA299N6.3, chromosome 20 open reading frame 201, hypothetical protein LOC198437, LKAAEAR motif containing 1, LKAAEAR1
Host Species Mouse
Immunogen Peptide Sequence - AIRLELFLPP (immunogen peptide is identical in mouse, rat, sheep, bovine and equine)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 198437
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.