Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LKB1/STK11 Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB337638 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB337638 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB337638 Supplier Novus Biologicals Supplier No. NBP214835APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LKB1/STK11 Polyclonal antibody specifically detects LKB1/STK11 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen LKB1/STK11
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate APC
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias EC 2.7.11.1, LKB1 serine/threonine kinase 11 (Peutz-Jeghers syndrome), PJS, PJS polarization-related protein LKB1, Renal carcinoma antigen NY-REN-19, serine/threonine kinase 11, serine/threonine-protein kinase 11, Serine/threonine-protein kinase LKB1, STK11
Host Species Rabbit
Immunogen A synthetic peptide made to a internal portion of the human LKB1 protein (between residues 250-350) [UniProt Q15831]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Hypoxia Signaling, mTOR Pathway, p53 Pathway, Phospho Specific, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 6794
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.