Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LKB1/STK11 Antibody (OTI0A3), FITC, Novus Biologicals™
SDP

Catalog No. NBP271300F Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP271300F 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP271300F Supplier Novus Biologicals Supplier No. NBP271300F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

LKB1/STK11 Monoclonal specifically detects LKB1/STK11 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.

Specifications

Antigen LKB1/STK11
Applications Immunohistochemistry
Classification Monoclonal
Clone OTI0A3
Conjugate FITC
Dilution Immunohistochemistry
Formulation PBS with 0.05% Sodium Azide
Gene Alias EC 2.7.11.1, LKB1 serine/threonine kinase 11 (Peutz-Jeghers syndrome), PJS, PJS polarization-related protein LKB1, Renal carcinoma antigen NY-REN-19, serine/threonine kinase 11, serine/threonine-protein kinase 11, Serine/threonine-protein kinase LKB1, STK11
Gene Symbols STK11
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 1-236 of human LKB1/STK11 (NP_000446) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Hypoxia Signaling, mTOR Pathway, p53 Pathway, Phospho Specific, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 6794.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.