Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LL-37 Antibody (B-L47), PerCP, Novus Biologicals™
SDP

Catalog No. NB340564 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB340564 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB340564 Supplier Novus Biologicals Supplier No. NBP318121PCP
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

LL-37 Monoclonal antibody specifically detects LL-37 in Human samples. It is validated for ELISA

Specifications

Antigen LL-37
Applications ELISA
Classification Monoclonal
Clone B-L47
Conjugate PerCP
Dilution ELISA
Formulation PBS
Gene Alias 18 kDa cationic antimicrobial protein, Antibacterial peptide FALL-39, CAP18, CAP-18, cathelicidin antimicrobial peptide, CRAMP, epididymis secretory sperm binding protein, FALL39, FALL-39, FALL-39 peptide antibiotic, hCAP-181, HSD26, LL37
Host Species Mouse
Immunogen Human antimicrobial peptide LL-37. Hybridoma: Myeloma X63/AG.8653 x Balb/c node cells
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 820
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.