Learn More
Invitrogen™ LLGL2 Polyclonal Antibody

Description
Immunogen sequence: RQVFVKCTLH PSDQLALEGP LSRVKSLKKS LRQSFRRMRR SRVSSHKRRP GGPTGEAQAQ AVNIKAERTG LQNMELAPVQ RKIEARSAED SFTGFVRTLY FADTYLRDSS RHCPSLWAGT NGGTVYAFSL RVPPAERRTD EPVRAEQAKE IQLMHRAPVV GILVLDGHNV PLPEPLEVAH DLSKSPDMQG SHQLLVVSEE.
Specifications
Specifications
| Antigen | LLGL2 |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 2.15 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | LLGL2 |
| Gene Accession No. | Q3TJ91, Q6P1M3 |
| Gene Alias | 9130006H11Rik; HGL; Hugl-2; human giant larvae homolog; lethal giant larvae homolog 2; lethal giant larvae homolog 2 (Drosophila); lethal giant larvae homolog 2, scribble cell polarity complex component; lethal giant larvae-like protein 2; Lethal(2) giant larvae protein homolog 2; LGL2; LLGL scribble cell polarity complex component 2; Llgl2; LLGL2, scribble cell polarity complex component; Llglh2; RGD1560307 |
| Gene Symbols | LLGL2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.