Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LMAN1 Antibody (OTI1A8) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NBP271152 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP271152 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP271152 Supplier Novus Biologicals Supplier No. NBP271152
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

LMAN1 Monoclonal specifically detects LMAN1 in Human, Mouse, Rat, Canine, Monkey, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, CyTOF-ready.

Specifications

Antigen LMAN1
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, CyTOF
Classification Monoclonal
Clone OTI1A8
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:200-500, Flow Cytometry 1:100, Immunohistochemistry 1:150, Immunocytochemistry/Immunofluorescence 1:100, CyTOF-ready
Formulation Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative
Gene Alias ERGIC-53, ERGIC53ER-Golgi intermediate compartment 53 kDa protein, F5F8Dendoplasmic reticulum-golgi intermediate compartment protein 53, FMFD1, Gp58, intracellular mannose specific lectin, Intracellular mannose-specific lectin MR60, Lectin mannose-binding 1, lectin, mannose-binding, 1, MCFD1coagulation factor V-factor VIII combined deficiency, MR60, protein ERGIC-53
Gene Symbols LMAN1
Host Species Mouse
Immunogen Full length human recombinant protein of human LMAN1 (NP_005561) produced in HEK293T cell.
Molecular Weight of Antigen 54.2 kDa
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 3998
Reconstitution Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended.
Target Species Human, Mouse, Rat, Canine, Monkey, Primate
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.