Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LMAN2 Antibody (02), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB328481 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB328481 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB328481 Supplier Novus Biologicals Supplier No. NBP305830APC
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

LMAN2 Monoclonal antibody specifically detects LMAN2 in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation

Specifications

Antigen LMAN2
Applications Western Blot, ELISA, Immunoprecipitation
Classification Monoclonal
Clone 02
Conjugate APC
Dilution Western Blot, ELISA, Immunoprecipitation
Formulation PBS
Gene Alias C5orf8, chromosome 5 open reading frame 8, Glycoprotein GP36b, GP36B, Lectin mannose-binding 2, lectin, mannose-binding 2, vesicular integral protein of 36 kDa, vesicular integral-membrane protein VIP36, VIP36
Host Species Mouse
Immunogen This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human LMAN2 (Uniprot#: Q12907; Asp 45-Arg 322).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10960
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.