Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LMO2 Antibody (1A9-3B11), HRP, Novus Biologicals™
SDP

Catalog No. NB11078626H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11078626H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB11078626H Supplier Novus Biologicals Supplier No. NB11078626H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

LMO2 Monoclonal specifically detects LMO2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen LMO2
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 1A9-3B11
Conjugate HRP
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Gene Accession No. P25791
Gene Alias Cysteine-rich protein TTG-2, LIM domain only 2 (rhombotin-like 1), LIM domain only protein 2, LMO-2, RBTN2rhombotin-2, RBTNL1rhombotin-like 1, RHOM2T-cell translocation protein 2, T-cell translocation gene 2, TTG2rhombotin 2
Gene Symbols LMO2
Host Species Mouse
Immunogen Recombinant human LMO2. [UniProt# P25791]
Molecular Weight of Antigen 24 kDa
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer
Primary or Secondary Primary
Gene ID (Entrez) 4005
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.