Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LMO2 Antibody (LMO2/3147R), DyLight 755, Novus Biologicals™
SDP

Catalog No. NB389164 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB389164 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB389164 Supplier Novus Biologicals Supplier No. NBP308945IR
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

LMO2 Monoclonal antibody specifically detects LMO2 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen LMO2
Applications Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone LMO2/3147R
Conjugate DyLight 755
Formulation 50mM Sodium Borate
Gene Alias Cysteine-rich protein TTG-2, LIM domain only 2 (rhombotin-like 1), LIM domain only protein 2, LMO-2, RBTN2rhombotin-2, RBTNL1rhombotin-like 1, RHOM2T-cell translocation protein 2, T-cell translocation gene 2, TTG2rhombotin 2
Host Species Rabbit
Immunogen Recombinant fragment (around aa 23-140) human LMO2 protein (exact sequence is proprietary) (Uniprot: P25791)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer
Primary or Secondary Primary
Gene ID (Entrez) 4005
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.