Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LMO7 Antibody, Novus Biologicals™
SDP

Catalog No. NB1001360 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1001360 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1001360 Supplier Novus Biologicals Supplier No. NB1001360
Add to Cart
Add to Cart

Goat Polyclonal Antibody

LMO7 Polyclonal specifically detects LMO7 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry (Frozen), Peptide ELISA, Western Blot.

Specifications

Antigen LMO7
Applications Immunohistochemistry, Immunohistochemistry (Frozen), Peptide ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5 - 2 ug/mL, Immunohistochemistry-Frozen, Peptide ELISA Detection limit 1:32000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. AAD33924.
Gene Alias FBX20F-box only protein 20, KIAA0858FBXO20F-box protein Fbx20, LIM domain 7, LIM domain only 7, LIM domain only protein 7, LMO-7, LOMP, zinc-finger domain-containing protein
Gene Symbols LMO7
Host Species Goat
Immunogen Peptide with sequence ESTCVSASLPRS corresponding to internal region according to AAD33924.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4008
Test Specificity Please note that the amino acid sequence of the immuning peptide differs one amino acid from the current NCBI accession numbers (NP_005349.3, NP_056667.2). This antibody is expected to recognize both reported isoforms (NP_005349.3; NP_056667.2).
Target Species Human, Mouse
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.