Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LMP2/PSMB9 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$480.74
Specifications
| Antigen | LMP2/PSMB9 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP158331
![]() |
Novus Biologicals
NBP158331 |
100 μL |
Each for $480.74
|
|
|||||
NBP1583320
![]() |
Novus Biologicals
NBP15833120UL |
20 μL | N/A | N/A | N/A | ||||
Description
LMP2/PSMB9 Polyclonal specifically detects LMP2/PSMB9 in Human samples. It is validated for Western Blot.Specifications
| LMP2/PSMB9 | |
| Polyclonal | |
| Rabbit | |
| P28065 | |
| 5698 | |
| IgG | |
| This product is specific to Subunit or Isoform: beta type-9. |
| Western Blot | |
| Unconjugated | |
| RUO | |
| beta1i, LMP2MGC70470, Low molecular mass protein 2, Macropain chain 7, Multicatalytic endopeptidase complex chain 7, proteasome (prosome, macropain) subunit, beta type, 9 (large multifunctionalpeptidase 2), proteasome catalytic subunit 1i, Proteasome chain 7, proteasome subunit beta 6i, proteasome subunit beta type-9, Proteasome subunit beta-1i, proteasome-related gene 2, PSMB6iproteasome (prosome, macropain) subunit, beta type, 9 (large multifunctionalprotease 2), Really interesting new gene 12 protein, RING12EC 3.4.25.1 | |
| PSMB9 | |
| Primary | |
| 21 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title