Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LMP2/PSMB9 Antibody (OTI1D1), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. N271261A700 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N271261A700 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N271261A700 Supplier Novus Biologicals Supplier No. NBP271261AF700
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

LMP2/PSMB9 Monoclonal specifically detects LMP2/PSMB9 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen LMP2/PSMB9
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI1D1
Conjugate Alexa Fluor 700
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias beta1i, LMP2MGC70470, Low molecular mass protein 2, Macropain chain 7, Multicatalytic endopeptidase complex chain 7, proteasome (prosome, macropain) subunit, beta type, 9 (large multifunctionalpeptidase 2), proteasome catalytic subunit 1i, Proteasome chain 7, proteasome subunit beta 6i, proteasome subunit beta type-9, Proteasome subunit beta-1i, proteasome-related gene 2, PSMB6iproteasome (prosome, macropain) subunit, beta type, 9 (large multifunctionalprotease 2), Really interesting new gene 12 protein, RING12EC 3.4.25.1
Gene Symbols PSMB9
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 21-219 of human LMP2/PSMB9 (NP_002791) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 5698.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.