Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LMTK2 Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB289854 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB289854 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB289854 Supplier Novus Biologicals Supplier No. NBP232182APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LMTK2 Polyclonal antibody specifically detects LMTK2 in Human samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen LMTK2
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate APC
Dilution Western Blot, Immunoprecipitation
Formulation PBS
Gene Alias AATYK2FLJ46659, Apoptosis-associated tyrosine kinase 2, Brain-enriched kinase, BREKCDK5/p35-regulated kinase, cprk, EC 2.7.11, KIAA1079hBREK, kinase phosphatase inhibitor 2, Kinase/phosphatase/inhibitor 2, KPI-2, KPI2EC 2.7.11.1, lemur tyrosine kinase 2Serine/threonine-protein kinase KPI-2, LMR2cyclin-dependent kinase 5/p35-regulated kinase, serine/threonine-protein kinase LMTK2
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 675 to 725 of human Lemur Tyrosine Kinase 2 using the numbering given in entry NP_055731.2
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 22853
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.