Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LMW E.Coli Host Cell Protein Antibody, Novus Biologicals™
SDP

Catalog No. NBP24420750 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP24420750 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP24420750 Supplier Novus Biologicals Supplier No. NBP24420750UG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LMW E.Coli Host Cell Protein Polyclonal specifically detects LMW E.Coli Host Cell Protein in E. coli samples. It is validated for Western Blot, ELISA.

Specifications

Antigen LMW E.Coli Host Cell Protein
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 1.29 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000, ELISA 1:1000-1:2000
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% sodium azide
Host Species Rabbit
Immunogen Anti-LMW E.Coli Host Cell Proteins Antibody was prepared by repeated immunizations with a mixture low molecular weight (LMW) proteins from E.Coli (BL21).
Quantity 50 μg
Primary or Secondary Primary
Target Species C. elegans, E. coli
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.