Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LMW E.Coli Host Cell Protein Antibody, Novus Biologicals™
SDP

Catalog No. NBP244207 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP244207 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP244207 Supplier Novus Biologicals Supplier No. NBP244207
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects LMW E. coli Host Cell Protein in C. elegans, E. coli samples, and it is validated for ELISA, Western Blotting

LMW E.Coli Host Cell Protein Polyclonal specifically detects LMW E.Coli Host Cell Protein in E. coli samples. It is validated for Western Blot, ELISA.

Specifications

Antigen LMW E.Coli Host Cell Protein
Applications Western Blot, ELISA, Dot Blot
Classification Polyclonal
Concentration 1.29 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000, ELISA 1:1000-1:2000, Dot Blot
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
Host Species Rabbit
Immunogen LMW E.Coli Host Cell Protein Antibody was prepared by repeated immunizations with a mixture low molecular weight (LMW) proteins from E.Coli (BL21).
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species E. coli
Content And Storage Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.