Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LMW-PTP/ACP1 Antibody (5C11), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB160536 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB160536 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB160536 Supplier Novus Biologicals Supplier No. NBP259404AF532
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

LMW-PTP/ACP1 Monoclonal antibody specifically detects LMW-PTP/ACP1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunofluorescence

Specifications

Antigen LMW-PTP/ACP1
Applications Western Blot, Flow Cytometry, ELISA, Immunofluorescence
Classification Monoclonal
Clone 5C11
Conjugate Alexa Fluor 532
Formulation 50mM Sodium Borate
Gene Alias acid phosphatase 1, soluble, acid phosphatase of erythrocyte, Adipocyte acid phosphatase, cytoplasmic phosphotyrosyl protein phosphatase, EC 3.1.3.2, EC 3.1.3.48, HAAP, LMW-PTP, LMW-PTPase, Low molecular weight cytosolic acid phosphatase, low molecular weight phosphotyrosine protein phosphatase, MGC111030, MGC3499, protein tyrosine phosphatase, Red cell acid phosphatase 1
Host Species Mouse
Immunogen Recombinant human LMW-PTP/ACP1 (1-158aa) purified from E. coli
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Phosphatase
Primary or Secondary Primary
Gene ID (Entrez) 52
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.