Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LMW-PTP/ACP1 Antibody - Isoform B, Novus Biologicals™
SDP

Catalog No. NB10056477T Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056477T 0.025 mg
NB10056477 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10056477T Supplier Novus Biologicals Supplier No. NB10056477SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LMW-PTP/ACP1 Polyclonal specifically detects LMW-PTP/ACP1 in Bovine, Human, Pig samples. It is validated for Western Blot.

Specifications

Antigen LMW-PTP/ACP1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-3 ug/ml
Formulation PBS containing 0.2% gelatin with 0.05% Sodium Azide
Gene Accession No. NP_009030
Gene Alias acid phosphatase 1, soluble, acid phosphatase of erythrocyte, Adipocyte acid phosphatase, cytoplasmic phosphotyrosyl protein phosphatase, EC 3.1.3.2, EC 3.1.3.48, HAAP, LMW-PTP, LMW-PTPase, Low molecular weight cytosolic acid phosphatase, low molecular weight phosphotyrosine protein phosphatase, MGC111030, MGC3499, protein tyrosine phosphatase, Red cell acid phosphatase 1
Gene Symbols ACP1
Host Species Rabbit
Immunogen A portion of amino acids 40-80 of human ACP1 was used as the immunogen.
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Protein Phosphatase
Primary or Secondary Primary
Gene ID (Entrez) 52
Test Specificity Reacts with Bovine, Human and Porcine. 93% sequence identity with mouse, 86% with canine and 80% with rat proteins.
Target Species Bovine, Pig, Human, Pig
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.