Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LMW-PTP/ACP1 Mouse anti-Human, Mouse, Clone: 5C11, Novus Biologicals™
SDP

Catalog No. NBP259404X Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP259404X 100 μL
NBP25940450 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP259404X Supplier Novus Biologicals Supplier No. NBP259404100UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

LMW-PTP/ACP1 Monoclonal specifically detects LMW-PTP/ACP1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen LMW-PTP/ACP1
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 5C11
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000, ELISA, Immunocytochemistry/Immunofluorescence 1:100
Formulation Liquid. In PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide
Gene Alias acid phosphatase 1, soluble, acid phosphatase of erythrocyte, Adipocyte acid phosphatase, cytoplasmic phosphotyrosyl protein phosphatase, EC 3.1.3.2, EC 3.1.3.48, HAAP, LMW-PTP, LMW-PTPase, Low molecular weight cytosolic acid phosphatase, low molecular weight phosphotyrosine protein phosphatase, MGC111030, MGC3499, protein tyrosine phosphatase, Red cell acid phosphatase 1
Gene Symbols ACP1
Host Species Mouse
Immunogen Recombinant human ACP1 (1-158aa) purified from E. coli
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Protein Phosphatase
Primary or Secondary Primary
Gene ID (Entrez) 52
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.