Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LMX1b Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP2411940.025MG
Description
LMX1b Polyclonal specifically detects LMX1b in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
LMX1b | |
Polyclonal | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
LMX1B | |
Antibody was raised against a 17 amino acid peptide near the carboxy terminus of human LMX1B. The immunogen is located within amino acids 280 - 330 of LMX1B. Amino Acid Squence: CRLGQGEPGPGQGLGQEV | |
Affinity Purified | |
RUO | |
Primary | |
At least three isoforms are known to exist. This antibody is predicted to not cross-react with LMX1A. | |
Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
1 mg/mL | |
Western Blot 1 - 2 μg/mL, ELISA | |
LIM homeobox transcription factor 1, beta, LIM homeobox transcription factor 1-beta, LIM/homeobox protein 1.2, LIM/homeobox protein LMX1B, LMX1.2, LMX-1.2, MGC138325, MGC142051, NPS1 | |
Rabbit | |
45 kDa | |
0.025 mg | |
Neuroscience | |
4010 | |
Human, Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction