Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LMX1b Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
$196.10 - $499.50
Specifications
Antigen | LMX1b |
---|---|
Concentration | 1 mg/mL |
Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
LMX1b Polyclonal specifically detects LMX1b in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
LMX1b | |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human, Mouse, Rat | |
LIM homeobox transcription factor 1, beta, LIM homeobox transcription factor 1-beta, LIM/homeobox protein 1.2, LIM/homeobox protein LMX1B, LMX1.2, LMX-1.2, MGC138325, MGC142051, NPS1 | |
LMX1B | |
IgG | |
Affinity Purified | |
45 kDa |
1 mg/mL | |
Polyclonal | |
Rabbit | |
Neuroscience | |
PBS with 0.02% Sodium Azide | |
4010 | |
Antibody was raised against a 17 amino acid peptide near the carboxy terminus of human LMX1B. The immunogen is located within amino acids 280 - 330 of LMX1B. Amino Acid Squence: CRLGQGEPGPGQGLGQEV | |
Primary | |
At least three isoforms are known to exist. This antibody is predicted to not cross-react with LMX1A. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title