Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LNK/SH2B3 Antibody, HRP, Novus Biologicals™
SDP

Catalog No. NB146777 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB146777 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB146777 Supplier Novus Biologicals Supplier No. NBP306063H
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LNK/SH2B3 Polyclonal specifically detects LNK/SH2B3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen LNK/SH2B3
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate HRP
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias IDDM20, LNKSH2B adapter protein 3, Lymphocyte adapter protein, Lymphocyte-specific adapter protein Lnk, SH2B adaptor protein 3, Signal transduction protein Lnk
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human LNK/SH2B3 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10019
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.