Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lnx1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP18051820UL
Description
Lnx1 Polyclonal specifically detects Lnx1 in Human samples. It is validated for Western Blot.Specifications
| Lnx1 | |
| Polyclonal | |
| Western Blot 1:1000 | |
| NP_116011 | |
| LNX1 | |
| Synthetic peptide directed towards the C terminal of human LNX1. Peptide sequence SHREWDLPIYVISVEPGGVISRDGRIKTGDILLNVDGVELTEVSRSEAVA. | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS and 2% Sucrose with 0.09% Sodium Azide | |
| EC 6.3.2.-, ligand of numb-protein X 1ligand of numb-protein X, LNXmulti-PDZ-domain-containing protein, E3 ubiquitin-protein ligase LNX, MPDZ, Numb-binding protein 1, PDZ domain-containing ring finger protein 2, PDZRN2E3 ubiquitin-protein ligase LNX | |
| Rabbit | |
| 70 kDa | |
| 20 μL | |
| Cancer, Hypoxia, Lipid and Metabolism, Zinc Finger | |
| 84708 | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction