Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lnx1 Antibody (OTI1H9), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. N272517A405 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N272517A405 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N272517A405 Supplier Novus Biologicals Supplier No. NBP272517AF405
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Lnx1 Monoclonal specifically detects Lnx1 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen Lnx1
Applications Western Blot
Classification Monoclonal
Clone OTI1H9
Conjugate Alexa Fluor 405
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias EC 6.3.2.-, ligand of numb-protein X 1ligand of numb-protein X, LNXmulti-PDZ-domain-containing protein, E3 ubiquitin-protein ligase LNX, MPDZ, Numb-binding protein 1, PDZ domain-containing ring finger protein 2, PDZRN2E3 ubiquitin-protein ligase LNX
Gene Symbols LNX1
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 260-528 of human Lnx1 (NP_116011) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cancer, Hypoxia, Lipid and Metabolism, Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 84708.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.