Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lnx1 Antibody (OTI1H9), PE, Novus Biologicals™
SDP

Catalog No. NB288086 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB288086 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB288086 Supplier Novus Biologicals Supplier No. NBP272517PE
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Lnx1 Monoclonal antibody specifically detects Lnx1 in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen Lnx1
Applications Western Blot
Classification Monoclonal
Clone OTI1H9
Conjugate PE
Dilution Western Blot
Formulation PBS
Gene Alias EC 6.3.2.-, ligand of numb-protein X 1ligand of numb-protein X, LNXmulti-PDZ-domain-containing protein, E3 ubiquitin-protein ligase LNX, MPDZ, Numb-binding protein 1, PDZ domain-containing ring finger protein 2, PDZRN2E3 ubiquitin-protein ligase LNX
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 260-528 of human LNX1 (NP_116011) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Hypoxia, Lipid and Metabolism, Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 84708
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.