Learn More
Description
Specifications
Specifications
| Antigen | LOC388813 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | DyLight 650 |
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin |
| Formulation | 50mM Sodium Borate |
| Gene Alias | LOC388813 uncharacterized protein ENSP00000383407-like |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with E. coli-derived Human LOC388813/ AF165138.7 fragment. |
| Purification Method | Antigen and protein A Affinity-purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
