Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LOC645015 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$464.00
Specifications
Antigen | LOC645015 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
LOC645015 Polyclonal specifically detects LOC645015 in Human samples. It is validated for Western Blot.Specifications
LOC645015 | |
Polyclonal | |
Rabbit | |
Human | |
asparagine-linked glycosylation 1-like 8, pseudogene, LOC645015 asparagine-linked glycosylation 1 homolog pseudogene | |
LOC645015 | |
IgG | |
Affinity Purified |
Western Blot | |
Unconjugated | |
RUO | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
645015 | |
Synthetic peptides corresponding to LOC645015(similar to mannosyltransferase) The peptide sequence was selected from the middle region of LOC645015. Peptide sequence LPSLVCVITGQGPLTEYYSRPIHQKHFQHIQVCNPWLEAEDYPLLLGSVD. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title