Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Anomalous homeobox protein Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP191410
Description
Anomalous homeobox protein Polyclonal specifically detects Anomalous homeobox protein in Human samples. It is validated for Western Blot.Specifications
| LOC647589 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| anomalous homeobox, hypothetical protein LOC647589 | |
| Rabbit | |
| 42 kDa | |
| 100 μL | |
| Primary | |
| This product recognizes Human. Dog: 86%; Horse: 79%. | |
| Human, Canine, Equine, Rabbit | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| NP_001177983 | |
| ANHX | |
| Synthetic peptide directed towards the n terminal of human hCG_2007354. Peptide sequence LRLHLLDNADVALACARVLDQQEQQQAACRLLEGCQVPGGSQELVQLWND. | |
| Affinity purified | |
| RUO | |
| 647589 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction