Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LOH12CR1 Antibody (OTI1E1), PE, Novus Biologicals™
SDP

Catalog No. NB288083 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB288083 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB288083 Supplier Novus Biologicals Supplier No. NBP272518PE
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

LOH12CR1 Monoclonal antibody specifically detects LOH12CR1 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen LOH12CR1
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone OTI1E1
Conjugate PE
Dilution Western Blot, Immunocytochemistry/ Immunofluorescence
Formulation PBS
Gene Alias LOH1CR12, loss of heterozygosity 12 chromosomal region 1 protein, loss of heterozygosity, 12, chromosomal region 1
Host Species Mouse
Immunogen Full length human recombinant protein of human LOH12CR1(NP_477517) produced in HEK293T cell.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 118426
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.