Learn More
LONP1 Rabbit anti-Human, Polyclonal, Bioss

Description
Specifications
Specifications
| Antigen | LONP1 |
| Applications | Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 μg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH |
| Gene | LONP1 |
| Gene Accession No. | P36776 |
| Gene Alias | 1200017E13Rik; CODASS; h LonHS; HAMAP-Rule:MF_03120}; hLON; hLON ATP-dependent protease; Lon; lon peptidase 1, mitochondrial; Lon protease homolog, mitochondrial; lon protease homolog, mitochondrial {ECO:0000255; Lon protease-like protein; lon protease-like protein {ECO:0000255; LONHs; LONP; LONP {ECO:0000255; Lonp1; Mitochondrial ATP-dependent protease Lon; mitochondrial ATP-dependent protease Lon {ECO:0000255; mitochondrial lon protease-like protein; PIM1; protease, serine, 15; PRSS15; Serine protease 15; serine protease 15 {ECO:0000255 |
| Gene Symbols | LONP1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.