Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LONRF2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP159292 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP159292 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP159292 Supplier Novus Biologicals Supplier No. NBP159292
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LONRF2 Polyclonal specifically detects LONRF2 in Human samples. It is validated for Western Blot.

Specifications

Antigen LONRF2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q1L5Z9
Gene Alias FLJ45273, LON peptidase N-terminal domain and ring finger 2, LON peptidase N-terminal domain and RING finger protein 2, MGC126711, Neuroblastoma apoptosis-related protease, RING finger protein 192, RNF192MGC126713
Gene Symbols LONRF2
Host Species Rabbit
Immunogen Synthetic peptides corresponding to LONRF2(LON peptidase N-terminal domain and ring finger 2) The peptide sequence was selected from the middle region of LONRF2. Peptide sequence FGMCLSAEHAGLSEYGCMLEIKDVRTFPDGSSVVDAIGISRFRVLSHRHR.
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 164832
Test Specificity Expected identity based on immunogen sequence: Canine: 100%; Human: 100%; Mouse: 100%; Rat: 100%; Equine: 78%.
Reconstitution Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Target Species Human, Mouse, Rat, Pig, Canine, Equine
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.