Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LONRF2 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$206.00 - $487.50

Specifications

Antigen LONRF2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP15929520
SDP
View Documents
Novus Biologicals
NBP15929520UL
20 μL
Each for $206.00
Only null left
Add to Cart
 
NBP159295
SDP
View Documents
Novus Biologicals
NBP159295
100 μL
Each for $487.50
Only null left
Add to Cart
 
Description

Description

LONRF2 Polyclonal specifically detects LONRF2 in Human samples. It is validated for Western Blot.
Specifications

Specifications

LONRF2
Polyclonal
Rabbit
Zinc Finger
FLJ45273, LON peptidase N-terminal domain and ring finger 2, LON peptidase N-terminal domain and RING finger protein 2, MGC126711, Neuroblastoma apoptosis-related protease, RING finger protein 192, RNF192MGC126713
LONRF2
IgG
Western Blot
Unconjugated
RUO
Q1L5Z9
164832
Synthetic peptides corresponding to LONRF2(LON peptidase N-terminal domain and ring finger 2) The peptide sequence was selected from the middle region of LONRF2. Peptide sequence IGISRFRVLSHRHRDGYNTADIEYLEDEKVEGPEYEELAALHDSVHQQSV.
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.