Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LONRF3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA529446
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIPA529446 100 μL
1 options

Catalog No. PIPA529446

Supplier: Invitrogen™ PA529446

Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: H1299, HepG2, Molt-4. Predicted reactivity: Bovine (82%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The protein encoded by this gene contains a RING finger domain, a motif present in a variety of functionally distinct proteins and known to be involved in protein-protein and protein-DNA interactions. Multiple alternatively spliced transcript variants have been suggested, but their full length natures are not clear.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LONRF3
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene LONRF3
Gene Accession No. Q496Y0
Gene Alias LON peptidase N-terminal domain and ring finger 3; LON peptidase N-terminal domain and RING finger protein 3; LONRF3; RING finger protein 127; RNF127
Gene Symbols LONRF3
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human LONRF3. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79836
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.