Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lonza™ Ficoll™ DNA Loading Buffer, 6X

Catalog No. bma50655
Change view
Click to view available options
Quantity:
5 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BMA50655 5 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BMA50655 Supplier Lonza™ Supplier No. 50655
Only null left
Add to Cart
Add to Cart

Ideal for improving resolution and separation of DNA fragments

  • Dark blue liquid
  • Soluble in water
  • Contains EDTA, xylene cyanol, and bromophenol blue

TRUSTED_SUSTAINABILITY

Specifications

Color Dark Blue
Concentration 6X
Form Liquid
Formulation Contains Ficoll™ EDTA, Xylene cyanol and Bromophenol Blue
For Use With (Application) Improving resolution and separation of DNA fragments
Quantity 5 x 1 mL
Storage Requirements Room Temperature
Product Type DNA Sample Loading Buffer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.