Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lonza™ FlashGel™ System Sample Preparation Reagents

Catalog No. bma57060
Click to view available options
Quantity:
2 x 500 μL
5 x 1 mL

For use with the Lonza FlashGel System. FlashGel™ System Sample Preparation Reagents for the FlashGel™ System provide the fastest separations and enable the user to watch as nucleic acids migrate in the gel.

  • Reagents include Recovery Buffer and Loading Dye
  • Loading Dye: 5X concentration; RNA native sample buffer

  • For research or manufacturing use only
  • Not for use in in vitro diagnostic procedures, drug use, or for administration to humans or animals

TRUSTED_SUSTAINABILITY

Specifications

Product Type DNA Recovery Buffer
Form Liquid
Quantity 2 x 500 μL
Storage Requirements Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.