Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lonza™ Formaldehyde Sample Buffer

Catalog No. bw50571
Click to view available options
Buffer:
Sample Buffer

Ideal when samples are to be recovered from agarose gels

  • For denaturation of RNA samples for RNA electrophoresis
  • Ready to use
  • Contains formaldehyde, formamide, and tracking dyes in MOPS buffer

TRUSTED_SUSTAINABILITY

Specifications

Buffer Sample Buffer
Product Type RNA Sample Loading Buffer-Formaldehyde
Form Liquid
Formulation Contains Formaldehyde, Formamide and Tracking dyes in MOPS Buffer
For Use With (Application) Denaturation of RNA samples for RNA electrophoresis when samples are to be recovered from Agarose gels
Quantity 5 x 1 mL
Storage Requirements −20°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.