Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LOX-1/OLR1 Antibody (214012) [DyLight 594], Novus Biologicals™
SDP

Catalog No. NB399483 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB399483 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB399483 Supplier Novus Biologicals Supplier No. FAB1564M
Add to Cart
Add to Cart

Rat Monoclonal Antibody

LOX-1/OLR1 Monoclonal antibody specifically detects LOX-1/OLR1 in Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF

Specifications

Antigen LOX-1/OLR1
Applications Western Blot, Flow Cytometry, CyTOF
Classification Monoclonal
Clone 214012
Conjugate DyLight 594
Dilution Western Blot, Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias CLEC8ASLOX1, C-type lectin domain family 8 member A, hLOX-1, Lectin-like oxidized LDL receptor 1, Lectin-like oxLDL receptor 1, Lectin-type oxidized LDL receptor 1, LOX-1, LOX1ox LDL receptor 1, LOXIN, oxidised low density lipoprotein (lectin-like) receptor 1, oxidized low density lipoprotein (lectin-like) receptor 1, oxidized low-density lipoprotein receptor 1, oxidized low-density lipoprotein receptor 1, soluble form, Ox-LDL receptor 1, SCARE1, scavenger receptor class E, member 1
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse LOX-1, Arg60-Ile363, Accession # AAG44998
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cardiovascular Biology, metabolism
Primary or Secondary Primary
Gene ID (Entrez) 4973
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.