Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LOX-1/OLR1 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB036719 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB036719 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB036719 Supplier Novus Biologicals Supplier No. NBP289136

Rabbit Polyclonal Antibody

LOX-1/OLR1 Polyclonal specifically detects LOX-1/OLR1 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen LOX-1/OLR1
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000, Flow Cytometry 1:50-1:100, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin
Formulation PBS (pH7.4), 0.2% BSA, 50% Glycerol with 0.05% Sodium Azide
Gene Alias CLEC8ASLOX1, C-type lectin domain family 8 member A, hLOX-1, Lectin-like oxidized LDL receptor 1, Lectin-like oxLDL receptor 1, Lectin-type oxidized LDL receptor 1, LOX-1, LOX1ox LDL receptor 1, LOXIN, oxidised low density lipoprotein (lectin-like) receptor 1, oxidized low density lipoprotein (lectin-like) receptor 1, oxidized low-density lipoprotein receptor 1, oxidized low-density lipoprotein receptor 1, soluble form, Ox-LDL receptor 1, SCARE1, scavenger receptor class E, member 1
Host Species Rabbit
Immunogen Recombinant protein within Human LOX-1/OLR1 aa 17-273/ 273. (SwissProt: P78380 Human; SwissProt: Q9EQ09 Mouse)
Purification Method Immunogen affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cardiovascular Biology, metabolism
Primary or Secondary Primary
Gene ID (Entrez) 4973
Target Species Human, Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.